Learn More
PLANT CELL TECHNOLOGY Broad-spectrum biocide for plant tissue culture, in-vitro laboratory use only, Plant Preservative Mixture™, PPM™, Clear Liquid, Anti-Contamination, 1L Bottle
Catalog No. NC3905173
Encompass_Preferred
Plant Preservative Mixture™ (PPM™) is a liquid additive designed to reduce microbial contamination in plant tissue culture and micropropagation applications. This 1L bottle provides ready-to-use solution that integrates easily into standard culture media. PPM™ contains broad-spectrum antimicrobial and antifungal components compatible with a wide variety of plant species, including ornamentals, woody plants, and edible crops. It supports aseptic culture and enhances explant survival rates in tissue culture applications. Packaged in a durable, clear 1L bottle with secure cap, PPM™ ensures safe handling and storage. Ideal for research laboratories, commercial propagation, and specialized plant cell culture applications. Stable under recommended storage conditions.
Catalog No. NC3905173 Supplier PLANT CELL TECHNOLOGY Supplier No. PPM1000
Plant Preservative Mixture™ (PPM™) is a liquid additive designed to reduce microbial contamination in plant tissue culture and micropropagation applications. This 1L bottle provides ready-to-use solution that integrates easily into standard culture media. PPM™ contains broad-spectrum antimicrobial and antifungal components compatible with a wide variety of plant species, including ornamentals, woody plants, and edible crops. It supports aseptic culture and enhances explant survival rates in tissue culture applications. Packaged in a durable, clear 1L bottle with secure cap, PPM™ ensures safe handling and storage. Ideal for research laboratories, commercial propagation, and specialized plant cell culture applications. Stable under recommended storage conditions.