Learn More
Weber Scientific BROAD SPECTRUM MICROTABS WEBER
SDP
Catalog No. NC0547003
Encompass
A superior tablet used for the preservation of milk samples to be analyzed on infrared instruments.
Catalog No. NC0547003 Supplier Weber Scientific Supplier No. 303850
A superior tablet used for the preservation of milk samples to be analyzed on infrared instruments.