Learn More
Broadline Medical Systems Lab Coat
Catalog No. 2210010E
Change view
Click to view available options
White; XX-Large
Catalog No.
22-100-10E White; XX-Large
1 options
This item is not returnable. View return policy
Catalog No. 22-100-10E Supplier Broadline Medical Systems Supplier No. B774213
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Round
Seam Type Serged