Learn More
eMolecules​ 943858-70-6 | Azidobutyric acid NHS ester | Broadpharm | MFCD22573606 | 226.192 | C8H10N4O4 | 95.000 | [N-]=[N+]=NCCCC(=O)ON1C(=O)CCC1=O | 500mg | 779523831
Catalog No. 5000126046 Shop All eMolecules Inc. Products
Encompass_Preferred
Azidobutyric acid NHS ester | Broadpharm | 943858-70-6 | MFCD22573606 | 226.192 | C8H10N4O4 | 95.000 | [N-]=[N+]=NCCCC(=O)ON1C(=O)CCC1=O | 500mg | 779523831
Catalog No. 50-001-26046 Supplier eMolecules​ Supplier No. 779523831
Azidobutyric acid NHS ester | Broadpharm | 943858-70-6 | MFCD22573606 | 226.192 | C8H10N4O4 | 95.000 | [N-]=[N+]=NCCCC(=O)ON1C(=O)CCC1=O | 500mg | 779523831