Learn More
Medchemexpress LLC Broflanilide | 1207727-04-5 | 99.0% | 663.28 | 100 MG
SDP
Catalog No. 5000344449
Encompass_Preferred
Broflanilide is an insecticide that converts into Desmethyl-Broflanilide, a powerful antagonist at the insect RDL GABA Receptor. This mechanism inhibits *S. litura* RDL GABAR with an IC50 of 1.3 nM. Desmethyl-Broflanilide shows limited activity against human GABAA α1β2γ2 receptor, mammalian GABAA α1β3γ2 receptor, and human glycine receptor (GlyR) α1β with IC50s greater than 3 μM, and less potent inhibition of GlyR α1-A288Gβ and GlyR α1-A288G (IC50s, 0.706, 0.149 μM).
  • Acts as a potent antagonist at the insect resistant-to-dieldrin (RDL) GABA Receptor.
  • Inhibits *S. litura* RDL GABAR.
  • Exhibits weak activity against human and mammalian GABAA receptors.
  • Less potently inhibits GlyR α1-A288Gβ and GlyR α1-A288G.
Catalog No. 50-003-44449 Supplier Medchemexpress LLC Supplier No. HY10868910MG
Broflanilide is an insecticide that converts into Desmethyl-Broflanilide, a powerful antagonist at the insect RDL GABA Receptor. This mechanism inhibits *S. litura* RDL GABAR with an IC50 of 1.3 nM. Desmethyl-Broflanilide shows limited activity against human GABAA α1β2γ2 receptor, mammalian GABAA α1β3γ2 receptor, and human glycine receptor (GlyR) α1β with IC50s greater than 3 μM, and less potent inhibition of GlyR α1-A288Gβ and GlyR α1-A288G (IC50s, 0.706, 0.149 μM).
  • Acts as a potent antagonist at the insect resistant-to-dieldrin (RDL) GABA Receptor.
  • Inhibits *S. litura* RDL GABAR.
  • Exhibits weak activity against human and mammalian GABAA receptors.
  • Less potently inhibits GlyR α1-A288Gβ and GlyR α1-A288G.