Learn More
Medchemexpress LLC Broflanilide | 1207727-04-5 | 99.0% | 663.28 | 2 MG
SDP
Catalog No. 5000345421
Encompass_Preferred
Broflanilide is a potential insecticide that metabolizes to Desmethyl-Broflanilide. Desmethyl-Broflanilide is a potent antagonist at the insect resistant-to-dieldrin (RDL) GABA Receptor, inhibiting S. litura RDL GABAR with an IC50 value of 1.3 nM.
  • It is a potential insecticide.
  • It metabolizes into an active form, Desmethyl-Broflanilide.
  • The metabolite, Desmethyl-Broflanilide, acts as a potent antagonist at the insect GABA Receptor.
  • The metabolite demonstrates selective inhibition against S. litura RDL GABAR, with an IC50 of 1.3 nM.
  • The metabolite exhibits weak activity against human and mammalian GABAA receptors and human glycine receptors.
Catalog No. 50-003-45421 Supplier Medchemexpress LLC Supplier No. HY108689100MG
Broflanilide is a potential insecticide that metabolizes to Desmethyl-Broflanilide. Desmethyl-Broflanilide is a potent antagonist at the insect resistant-to-dieldrin (RDL) GABA Receptor, inhibiting S. litura RDL GABAR with an IC50 value of 1.3 nM.
  • It is a potential insecticide.
  • It metabolizes into an active form, Desmethyl-Broflanilide.
  • The metabolite, Desmethyl-Broflanilide, acts as a potent antagonist at the insect GABA Receptor.
  • The metabolite demonstrates selective inhibition against S. litura RDL GABAR, with an IC50 of 1.3 nM.
  • The metabolite exhibits weak activity against human and mammalian GABAA receptors and human glycine receptors.