Learn More
Sigma Aldrich Fine Chemicals Biosciences Broken-glass disposal boxe
Catalog No. 502900459
Encompass
Corrugated cardboard receptacle supplied with 2 mil PE bag. When full safety cap is pulled into place and entire unit is ready for disposal. Printed in four languages (English french Spanish and German). Shipped flat.
Catalog No. 50-290-0459 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2071871PAK
Corrugated cardboard receptacle supplied with 2 mil PE bag. When full safety cap is pulled into place and entire unit is ready for disposal. Printed in four languages (English french Spanish and German). Shipped flat.