Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromanilic Acid 98.0+%, TCI America™
SDP

Catalog No. D018210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D018210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D018210G Supplier TCI America Supplier No. D018210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4370-59-6
Molecular Formula C6H2Br2O4
Molecular Weight (g/mol) 297.89
MDL Number MFCD00058956
InChI Key GMZWPTALVQRAFV-UHFFFAOYSA-N
Synonym 2,5-Dibromo-3,6-dihydroxy-p-quinone
PubChem CID 78078
IUPAC Name 2,5-dibromo-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES OC1=C(Br)C(=O)C(O)=C(Br)C1=O

Specifications

Melting Point 270°C
Quantity 10 g
Formula Weight 297.89
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bromanilic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.