Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromate-Bromide, For Phenols, Certified, 0.100N ±0.001N (0.017M), LabChem™

Catalog No. LC119504
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC119504 4 L
LC119501 500 mL
LC119502 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC119504 Supplier LabChem, Inc. Supplier No. LC119504
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Bromate-Bromide
Color Colorless
Name Note For Phenols
CAS Max % 98.72, 1, 0.28
CAS 7758-01-2
Linear Formula KBrO3
Concentration 0.100N ±0.001N (0.017M)
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 167
Grade Certified
Identification Passes Test
Packaging Amber Glass
Traceability to NIST Traceable to NIST
Decomposition Information Hydrogen bromide
Quantity 4 L
Physical Form Liquid
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.