Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromate Standard, Ultra Scientific

Catalog No. USICC010
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICC010 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICC010 Supplier Agilent Technologies Supplier No. ICC010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5

Specifications

Density 1
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material Bromate Standard, 1 analyte at 1000μg/mL in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.