Learn More
Medchemexpress LLC Brombuterol-d9 | 1353940-55-2 | 99.0% | 1 MG
SDP
Catalog No. 5000338649
Encompass_Preferred
Brombuterol-d9 is a deuterium labeled Brombuterol. Brombuterol is a β-adrenergic receptor agonist.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets β adrenergic receptor
Catalog No. 50-003-38649 Supplier Medchemexpress LLC Supplier No. HY131104S1MG
Brombuterol-d9 is a deuterium labeled Brombuterol. Brombuterol is a β-adrenergic receptor agonist.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Targets β adrenergic receptor