Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Brombuterol hydrochloride | 21912-49-2 | MFCD10567368 | 99.9% | 402.55 g·mol⁻1 | C12H19Br2ClN2O | 5 MG
SDP

Catalog No. 5000262944
Encompass_Preferred

Brombuterol hydrochloride is a β-adrenergic receptor agonist provided as an analytical research standard for laboratory use. It is intended for research purposes and not for human consumption.

  • High purity: 99.9%.
  • Small pack sizes suitable for analytical assays.
  • Molecular weight 402.55 g·mol⁻1.
  • CAS number 21912-49-2 for unambiguous identification.
  • Store sealed at 4°C; in solution keep at -80°C for long-term storage.

Catalog No. 50-002-62944 Supplier Medchemexpress LLC Supplier No. HY1311455MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brombuterol hydrochloride is a β-adrenergic receptor agonist provided as an analytical research standard for laboratory use. It is intended for research purposes and not for human consumption.

  • High purity: 99.9%.
  • Small pack sizes suitable for analytical assays.
  • Molecular weight 402.55 g·mol⁻1.
  • CAS number 21912-49-2 for unambiguous identification.
  • Store sealed at 4°C; in solution keep at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.