Learn More
Hach Company Bromcresol Green-Methyl Red Indicator Powder Pillows, pk/100
Catalog No. 501994475 Shop All Hach Company Products
Encompass
pH indicator for determination of total alkalinity by titration methods. Pack of 100 Powder Pillows
Catalog No. 50-199-4475 Supplier Hach Company Supplier No. 94399
pH indicator for determination of total alkalinity by titration methods. Pack of 100 Powder Pillows