Learn More
Hach Company Bromcresol Green-Methyl Red Indicator Powder, 15 g
Supplier:  Hach Company 94323
Encompass
pH indicator powder for determination of total alkalinity by titration. 15 g bottle.
Catalog No. 50-199-4474