Learn More
Hach Company Bromcresol Green Indicator Solution, 500 mL
Catalog No. 501995819 Shop All Hach Company Products
Encompass
pH indicator: pH 3.8 (yellow)-pH 5.4 (blue). 500 mL Bottle
Catalog No. 50-199-5819 Supplier Hach Company Supplier No. 2329549
pH indicator: pH 3.8 (yellow)-pH 5.4 (blue). 500 mL Bottle