Learn More
Hach Company Bromcresol Green-Methyl Red Indicator Solution, 500 mL
Supplier:  Hach Company 2329249
Encompass
pH indicator for alkalinity determination by titration. APHA formulation 500 mL Bottle
Catalog No. 50-199-5816