Learn More
Crescent Chemical Co Inc BROMELAIN FROM PINEAPPLE
SDP
Supplier:  Crescent Chemical Co Inc SE15250.02
Encompass_Preferred
Bromelain from pineapple stem min. 0.3 DMC-U/mg powder Suitable for blood group serology (1) and degradation of proteins. Size - 100G Storage Conditions - +2 °C TO +8 °C Catalog Number - 15250.02 CAS 37189-34-7
Catalog No. 50-247-379