Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromelain, MP Biomedicals™

Catalog No. ICN10114291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10114291 1 kg
ICN10114210 10 g
ICN10114225 25 g
ICN10114290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10114291 Supplier MP Biomedicals, Inc Supplier No. 0210114291
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 9001-00-7

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-00-7

Specifications

Color Tan to White-Yellow
Quantity 1 kg
Physical Form Powder
Chemical Name or Material Bromelain
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.