Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromelain from Pineapple, MP Biomedicals

Catalog No. MP210114280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210114280 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210114280 Supplier MP Biomedicals, Inc Supplier No. 0210114280
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Bromelain may be used to inhibit the biosysnthesis of proinflammatory prostaglandins
  • It may be used to reduce clotting efficiency
  • Bromelain, from pineapple stem, has been used to make enzymatic hydrolysates of honeybee-collected pollen

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-00-7
MDL Number MFCD00081423

Specifications

Color Tan
Quantity 100 g
Solubility Information Readily soluble in water, insoluble in most organic solvents such as acetone, ether, ethanol and methanol.
Formula Weight Approximately 33,000. (Lit.)
Physical Form Powder
Chemical Name or Material BROMELAIN from Pineapple
Recommended Storage -20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.