Learn More
Cayman Chemical Bromfenac-d4sodIum salt 500ug
SDP
Catalog No. 502935235
Encompass_Preferred
An internal standard for the quantification of bromfenac by GC- or LC-MS
Catalog No. 50-293-5235 Supplier Cayman Chemical Supplier No. 26598500 UG
An internal standard for the quantification of bromfenac by GC- or LC-MS