Learn More
Medchemexpress LLC Bromfenac sodium | 91714-93-1 | 99.9% | 200 MG
SDP
Catalog No. 5000410647
Encompass_Preferred
Bromfenac sodium is a potent and orally active inhibitor of COX, with IC50s of 5.56 and 7.45 nM for COX-1 and COX-2, respectively. It can be used in ocular inflammation research.
  • Potent and orally active inhibitor of COX
  • IC50s of 5.56 nM for COX-1 and 7.45 nM for COX-2
  • Can be used in ocular inflammation research
  • Solid appearance
  • Light yellow to yellow color
Catalog No. 50-004-10647 Supplier Medchemexpress LLC Supplier No. HYB1888A200MG
Bromfenac sodium is a potent and orally active inhibitor of COX, with IC50s of 5.56 and 7.45 nM for COX-1 and COX-2, respectively. It can be used in ocular inflammation research.
  • Potent and orally active inhibitor of COX
  • IC50s of 5.56 nM for COX-1 and 7.45 nM for COX-2
  • Can be used in ocular inflammation research
  • Solid appearance
  • Light yellow to yellow color