Learn More
Medchemexpress LLC Bromfenac (sodium) | 91714-93-1 | MFCD09953182 | 99.9% | 356.15 | C15H11BrNNaO3 | 10 MG
SDP
Catalog No. 5000299010
Encompass_Preferred
Bromfenac (sodium) is the monosodium salt of bromfenac, a brominated non-steroidal anti-inflammatory research reagent that potently inhibits cyclooxygenase enzymes. It is intended for use in studies of COX-mediated inflammation, including ocular inflammation models.
  • Potent COX-1 and COX-2 inhibitor (IC50: 5.56 nM and 7.45 nM).
  • High purity research-grade material (99.9% reported purity).
  • Anhydrous sodium salt with molecular weight 356.15 g/mol.
  • Available in small research pack sizes suitable for screening and experimental use.
  • Useful for in vitro and in vivo inflammation studies and ocular research.
  • CAS number 91714-93-1 for unambiguous identification.
Catalog No. 50-002-99010 Supplier Medchemexpress LLC Supplier No. HYB1888A10MG
Bromfenac (sodium) is the monosodium salt of bromfenac, a brominated non-steroidal anti-inflammatory research reagent that potently inhibits cyclooxygenase enzymes. It is intended for use in studies of COX-mediated inflammation, including ocular inflammation models.
  • Potent COX-1 and COX-2 inhibitor (IC50: 5.56 nM and 7.45 nM).
  • High purity research-grade material (99.9% reported purity).
  • Anhydrous sodium salt with molecular weight 356.15 g/mol.
  • Available in small research pack sizes suitable for screening and experimental use.
  • Useful for in vitro and in vivo inflammation studies and ocular research.
  • CAS number 91714-93-1 for unambiguous identification.