Learn More
Medchemexpress LLC Bromfenac sodium hydrate | 120638-55-3 | 99.9% | 200 MG
SDP
Catalog No. 5000409794
Encompass_Preferred
Bromfenac sodium hydrate is a potent and orally active inhibitor of COX, with IC50s of 5.56 and 7.45 nM for COX-1 and COX-2, respectively. It can be used in ocular inflammation research.
  • Potent and orally active inhibitor of COX.
  • Can be used in ocular inflammation research.
  • Available in solid and solution forms.
Catalog No. 50-004-09794 Supplier Medchemexpress LLC Supplier No. HYB1888B200MG
Bromfenac sodium hydrate is a potent and orally active inhibitor of COX, with IC50s of 5.56 and 7.45 nM for COX-1 and COX-2, respectively. It can be used in ocular inflammation research.
  • Potent and orally active inhibitor of COX.
  • Can be used in ocular inflammation research.
  • Available in solid and solution forms.