Learn More
Cayman Chemical Bromfenac (sodium salt), Purity: ≥99%, CAS: 91714-93-1, 50 mg
SDP
Catalog No. 502964477
Encompass_Preferred
A COX-2 inhibitor (IC50 = 6.6 nM); selective over COX-1 (IC50 = 210 nM); inhibits PGE2 production in a rabbit model of LPS-induced eye inflammation; decreases inflammation following cataract removal in dogs (0.9% topically on a tapering schedule for 24 weeks)
Catalog No. 50-296-4477 Supplier Cayman Chemical Supplier No. 2376350 MG
A COX-2 inhibitor (IC50 = 6.6 nM); selective over COX-1 (IC50 = 210 nM); inhibits PGE2 production in a rabbit model of LPS-induced eye inflammation; decreases inflammation following cataract removal in dogs (0.9% topically on a tapering schedule for 24 weeks)