Learn More
Medchemexpress LLC Bromhexine-d3 hydrochloride | 611-75-6 | 99.0% | 1 MG
SDP
Catalog No. 5000407281
Encompass_Preferred
Bromhexine-d3 (hydrochloride) is a deuterium-labeled Bromhexine (hydrochloride) that functions as a potent and specific TMPRSS2 protease inhibitor with an IC50 of 0.75 μM. It holds potential for preventing and managing SARS-CoV-2 infection and also acts as an autophagy agonist. Additionally, Bromhexine hydrochloride serves as a mucolytic cough suppressant, suggesting its utility in various respiratory conditions. This product is intended for research use only.
  • Deuterium labeled Bromhexine (hydrochloride)
  • Potent and specific TMPRSS2 protease inhibitor
  • Can prevent and manage SARS-CoV-2 infection
  • Autophagy agonist
  • Mucolytic cough suppressant
  • Used as a tracer or internal standard for quantitative analysis
Catalog No. 50-004-07281 Supplier Medchemexpress LLC Supplier No. HYB0372AS1MG
Bromhexine-d3 (hydrochloride) is a deuterium-labeled Bromhexine (hydrochloride) that functions as a potent and specific TMPRSS2 protease inhibitor with an IC50 of 0.75 μM. It holds potential for preventing and managing SARS-CoV-2 infection and also acts as an autophagy agonist. Additionally, Bromhexine hydrochloride serves as a mucolytic cough suppressant, suggesting its utility in various respiratory conditions. This product is intended for research use only.
  • Deuterium labeled Bromhexine (hydrochloride)
  • Potent and specific TMPRSS2 protease inhibitor
  • Can prevent and manage SARS-CoV-2 infection
  • Autophagy agonist
  • Mucolytic cough suppressant
  • Used as a tracer or internal standard for quantitative analysis