Learn More
Medchemexpress LLC Bromhexine hydrochloride | 611-75-6 | 99.9% | 5 G
SDP
Catalog No. 5000406297
Encompass_Preferred
Bromhexine hydrochloride is a potent and specific TMPRSS2 protease inhibitor with an IC50 of 0.75 μM, which can help prevent and manage SARS-CoV-2 infection. It also functions as an autophagy agonist and a mucolytic cough suppressant, offering potential benefits for various respiratory conditions.
  • Potent and specific TMPRSS2 protease inhibitor
  • Helps prevent and manage SARS-CoV-2 infection
  • Autophagy agonist
  • Mucolytic cough suppressant
  • Potential for various respiratory conditions
Catalog No. 50-004-06297 Supplier Medchemexpress LLC Supplier No. HYB0372A5G
Bromhexine hydrochloride is a potent and specific TMPRSS2 protease inhibitor with an IC50 of 0.75 μM, which can help prevent and manage SARS-CoV-2 infection. It also functions as an autophagy agonist and a mucolytic cough suppressant, offering potential benefits for various respiratory conditions.
  • Potent and specific TMPRSS2 protease inhibitor
  • Helps prevent and manage SARS-CoV-2 infection
  • Autophagy agonist
  • Mucolytic cough suppressant
  • Potential for various respiratory conditions