Learn More
AccuStandard, Inc BROMIDE 100ML
SDP
Catalog No. NC9835194
Encompass
Anion Standard - Bromide. 100 mL. 1000 ?g/mL in 2% Sodium hydroxide.
Catalog No. NC9835194 Supplier AccuStandard, Inc Supplier No. ICBR1X1
Anion Standard - Bromide. 100 mL. 1000 ?g/mL in 2% Sodium hydroxide.