Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Bromide-Bromate 0.02 Normal NIST Traceable Solution ASTM D1159 (1L)
SDP

Catalog No. NC9984269
Encompass_Preferred

Bromide-Bromate 0.02 Normal NIST Traceable Solution ASTM D1159 (1L)

Catalog No. NC9984269 Supplier Aqua Solutions Supplier No. 13611L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromide-Bromate 0.02 Normal NIST Traceable Solution ASTM D1159 (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.