Learn More
ERA WATERS CORP BROMIDE QC
SDP
Catalog No. NC0765652
Encompass
One 15 mL screw-cap vial yields up to 2 liters after dilution. Use with ion chromatography or colorimetric methods.
Catalog No. NC0765652 Supplier ERA WATERS CORP Supplier No. 769
One 15 mL screw-cap vial yields up to 2 liters after dilution. Use with ion chromatography or colorimetric methods.