Learn More
SPEX CERTIPREP LLC 3P BROMIDE STD 1000PPM 250ML
Catalog No. NC0211466
Encompass_Preferred
(HPS by Zeptometrix) 1000 ug/mL Bromide in H2O; Single Component IC standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC0211466 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICBRM250
(HPS by Zeptometrix) 1000 ug/mL Bromide in H2O; Single Component IC standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.