Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromine (Bromide-Bromate), 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02002462 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-002-462 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-002-462 Supplier Avantor Supplier No. 562502
Add to Cart
Add to Cart

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.00kg
Quantity 1 L
Formula Weight 79.9
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Bromine (Bromide-Bromate), 0.1N Volumetric Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.