Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromine (Certified ACS), Fisher Chemical

Catalog No. B38550 Shop All Fisherbrand Products
Change view
Click to view available options
Amber Glass; 50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B385-50 Amber Glass; 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B385-50

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 58.7°C
Melting Point -7.2°C
Color Red Brown
CAS 7726-95-6
Physical Form Liquid
Quantity 50 mL
Assay Percent Range ≥99.5%
Molecular Formula Br
MDL Number MFCD00010896
UN Number 1744
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES [Br-]
IUPAC Name bromide
Molecular Weight (g/mol) 79.91
PubChem CID 24408
ChEBI CHEBI:29224
Formula Weight 159.81
Percent Purity ≥99.5%
Grade Certified ACS
Identification Pass Test
Packaging Amber Glass
Vapor Pressure 230mbar at 20°C
Density 3.111g/cm³
Chemical Name or Material Bromine
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.