Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromo[[1,3-bis[(4S,5S)-1-benzoyl-4,5-diphenyl-2-imidazolin-2-yl]benzene]palladium(II)], TCI America™
SDP

Catalog No. B3553100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3553100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3553100MG Supplier TCI America Supplier No. B3553100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1242081-29-3
Molecular Formula C50H37BrN4O2Pd
Molecular Weight (g/mol) 912.20
MDL Number MFCD16295192
InChI Key KZAGTVGVDHDPDZ-UHFFFAOYNA-M
Synonym [Bz-Phebim]Pd-Br
PubChem CID 131674829
IUPAC Name 2,6-bis(1-benzoyl-4,5-diphenyl-4,5-dihydro-1H-imidazol-2-yl)benzen-1-ide; bromopalladiumylium
SMILES Br[Pd+].O=C(N1C(C(N=C1C1=CC=CC(=[C-]1)C1=NC(C(N1C(=O)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 912.20
Physical Form Crystalline Powder
Chemical Name or Material Bromo[[1,3-bis[(4S,5S)-1-benzoyl-4,5-diphenyl-2-imidazolin-2-yl]benzene]palladium(II)]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.