Learn More
Crescent Chemical Co Inc BROMO-4-CHLORO-3-INDOLYL-B-D-G
SDP
Supplier:  Crescent Chemical Co Inc SE15243.03
Encompass_Preferred
Bromo-4-chloro-3-indolyl-b-D-galactoside (X-Gal) research grade X-Gal is a well known histochemical substrate used to detect the β-galactosidase enzyme (1, 2, 3, 4, 9, 10). Identification of lacf-colonies (8). X-gal is used to distinguish recombinant plasmids from parental vectors in cloning exp
Catalog No. 50-247-371