Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromo-PEG1-acid | 1393330-33-0 | 98.0% | 250 MG
SDP

Catalog No. 5000341455
Encompass_Preferred

Bromo-PEG1-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules contain two different ligands connected by a linker, facilitating the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Molecular weight: 197.03
  • Chemical formula: C5H9BrO3
  • Appearance: Liquid, light yellow to yellow

Catalog No. 50-003-41455 Supplier Medchemexpress LLC Supplier No. 250MGHY130425
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bromo-PEG1-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These molecules contain two different ligands connected by a linker, facilitating the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Molecular weight: 197.03
  • Chemical formula: C5H9BrO3
  • Appearance: Liquid, light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.