Learn More
Medchemexpress LLC Bromo-PEG1-C2-azide | 1144106-65-9 | 99.5% | C4H8BrN3O | 100 MG
SDP
Catalog No. 5000356657
Encompass_Preferred
Bromo-PEG1-C2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Click chemistry reagent with Azide group
  • Reacts with Alkyne groups via CuAAc
  • Reacts with DBCO or BCN groups via SPAAC
Catalog No. 50-003-56657 Supplier Medchemexpress LLC Supplier No. HY140819100MG
Bromo-PEG1-C2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Click chemistry reagent with Azide group
  • Reacts with Alkyne groups via CuAAc
  • Reacts with DBCO or BCN groups via SPAAC