Learn More
Medchemexpress LLC Bromo-PEG1-C2-azide | 1144106-65-9 | 99.5% | C4H8BrN3O | 25MG
SDP
Catalog No. 5000355958
Encompass_Preferred
Bromo-PEG1-C2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-003-55958 Supplier Medchemexpress LLC Supplier No. 25MGHY140819
Bromo-PEG1-C2-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilizes the intracellular ubiquitin-proteasome system