Learn More
Medchemexpress LLC Bromo-PEG1-C2-Boc | 1393330-36-3 | 98.58% | C9H17BrO3 | 250 MG
SDP
Catalog No. 5000352522
Encompass_Preferred
Bromo-PEG1-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. These compounds contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Contains two different ligands connected by a linker
  • Ligand for an E3 ubiquitin ligase
  • Ligand for target protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52522 Supplier Medchemexpress LLC Supplier No. 250MGHY141364
Bromo-PEG1-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. These compounds contain two different ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Contains two different ligands connected by a linker
  • Ligand for an E3 ubiquitin ligase
  • Ligand for target protein
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins