Learn More
Medchemexpress LLC Bromo-PEG1-CH2-Boc | 157759-50-7 | ≥98.0% | C8H15BrO3 | 250 MG
SDP
Catalog No. 5000352669
Encompass_Preferred
Bromo-PEG1-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Can be used in the synthesis of PROTACs
Catalog No. 50-003-52669 Supplier Medchemexpress LLC Supplier No. 250MGHY141370
Bromo-PEG1-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker
  • Can be used in the synthesis of PROTACs