Learn More
Medchemexpress LLC Bromo-PEG1-CH2-Boc | 157759-50-7 | >98% | C8H15BrO3 | 10 G
SDP
Catalog No. 5000356308
Encompass_Preferred
Bromo-PEG1-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase and the other for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56308 Supplier Medchemexpress LLC Supplier No. HY14137010G
Bromo-PEG1-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase and the other for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins