Learn More
Medchemexpress LLC Bromo-PEG2-C2-azide | 530151-56-5 | 98.7% | 1 G
SDP
Catalog No. 5000339959
Encompass_Preferred
Bromo-PEG2-C2-azide is a versatile non-cleavable linker utilized in the synthesis of antibody-drug conjugates (ADCs) and as a PEG-based PROTAC linker for PROTAC synthesis. This click chemistry reagent contains an azide group, enabling its use in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Non-cleavable ADC linker
  • PEG-based PROTAC linker
  • Contains an azide group for click chemistry
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Useful in synthesis of antibody-drug conjugates (ADCs)
  • Applicable in synthesis of PROTACs
Catalog No. 50-003-39959 Supplier Medchemexpress LLC Supplier No. HY1304851G
Bromo-PEG2-C2-azide is a versatile non-cleavable linker utilized in the synthesis of antibody-drug conjugates (ADCs) and as a PEG-based PROTAC linker for PROTAC synthesis. This click chemistry reagent contains an azide group, enabling its use in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Non-cleavable ADC linker
  • PEG-based PROTAC linker
  • Contains an azide group for click chemistry
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Useful in synthesis of antibody-drug conjugates (ADCs)
  • Applicable in synthesis of PROTACs