Learn More
Medchemexpress LLC Bromo-PEG2-C2-azide | 530151-56-5 | MFCD25424115 | 98.7% | 238.08 g·mol⁻1 | C6H12BrN3O2 | 100 MG
SDP
Catalog No. 5000202314
Encompass_Preferred
Bromo-PEG2-C2-azide is a PEG-based bromo azide linker used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PROTAC linker. It contains an azide functional group for click chemistry and a bromoalkyl handle for further functionalization. Molecular formula C6H12BrN3O2; molecular weight 238.08 g·mol⁻1; purity ≥98.7%.
  • Contains an azide group for copper-catalyzed and strain-promoted click reactions.
  • Provides a bromoalkyl handle for alkylation or substitution reactions.
  • Suitable as a non-cleavable linker for ADC synthesis and as a PROTAC linker.
  • High purity for reliable synthetic performance.
  • Available in small lab quantities for research use.
Catalog No. 50-002-02314 Supplier Medchemexpress LLC Supplier No. HY130485100MG
Bromo-PEG2-C2-azide is a PEG-based bromo azide linker used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as a PROTAC linker. It contains an azide functional group for click chemistry and a bromoalkyl handle for further functionalization. Molecular formula C6H12BrN3O2; molecular weight 238.08 g·mol⁻1; purity ≥98.7%.
  • Contains an azide group for copper-catalyzed and strain-promoted click reactions.
  • Provides a bromoalkyl handle for alkylation or substitution reactions.
  • Suitable as a non-cleavable linker for ADC synthesis and as a PROTAC linker.
  • High purity for reliable synthetic performance.
  • Available in small lab quantities for research use.