Learn More
Medchemexpress LLC Bromo-PEG2-C2-Boc | 1381861-91-1 | 97.0% | C11H21BrO4 | 10 G
SDP
Catalog No. 5000353105
Encompass_Preferred
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein.
  • PEG-based linker for PROTAC synthesis
  • Utilizes the ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
Catalog No. 50-003-53105 Supplier Medchemexpress LLC Supplier No. HY14136510G
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein.
  • PEG-based linker for PROTAC synthesis
  • Utilizes the ubiquitin-proteasome system
  • Designed for selective degradation of target proteins