Learn More
Medchemexpress LLC Bromo-PEG2-C2-Boc | 1381861-91-1 | 97.0% | C11H21BrO4 | 250 MG
SDP
Catalog No. 5000352648
Encompass_Preferred
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is for research use only and exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52648 Supplier Medchemexpress LLC Supplier No. 250MGHY141365
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is for research use only and exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins