Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromo-PEG2-C2-Boc | 1381861-91-1 | 97.0% | C11H21BrO4 | 50 G
SDP

Catalog No. 5000353862
Encompass_Preferred

Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker, catalog number HY-141365, designed for the synthesis of PROTACs. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker like this product. It is intended for research use only.

  • Purity: 97.0%
  • Molecular weight: 297.19
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Solubility: Soluble in DMSO (100 mg/mL)

Catalog No. 50-003-53862 Supplier Medchemexpress LLC Supplier No. HY14136550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker, catalog number HY-141365, designed for the synthesis of PROTACs. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker like this product. It is intended for research use only.

  • Purity: 97.0%
  • Molecular weight: 297.19
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Solubility: Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.