Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromo-PEG2-CH2-Boc | 1807518-63-3 | 97.0% | C10H19BrO4 | 1 G
SDP

Catalog No. 5000353214
Encompass_Preferred

Bromo-PEG2-CH2-Boc is a chemical compound with the formula C10H19BrO4 and a molecular weight of 283.16. It is commonly used as a laboratory chemical and for the manufacture of various substances, offering a high purity of 97.0%.

  • Appearance: Colorless to light yellow liquid
  • Suitable for laboratory chemical applications
  • Ideal for the manufacture of substances
  • Storage: Pure form stable at -20°C for 3 years or 4°C for 2 years

Catalog No. 50-003-53214 Supplier Medchemexpress LLC Supplier No. HY1413711G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromo-PEG2-CH2-Boc is a chemical compound with the formula C10H19BrO4 and a molecular weight of 283.16. It is commonly used as a laboratory chemical and for the manufacture of various substances, offering a high purity of 97.0%.

  • Appearance: Colorless to light yellow liquid
  • Suitable for laboratory chemical applications
  • Ideal for the manufacture of substances
  • Storage: Pure form stable at -20°C for 3 years or 4°C for 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.