Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromo-PEG2-CH2-Boc | 1807518-63-3 | 97.0% | C10H19BrO4 | 250 MG
SDP

Catalog No. 5000352670
Encompass_Preferred

Bromo-PEG2-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective protein degradation
  • Suitable for laboratory chemical applications
  • Purity of 97.0% (NMR)
  • Appearance: Colorless to light yellow liquid
  • Molecular weight: 283.16
  • Chemical formula: C10H19BrO4
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-52670 Supplier Medchemexpress LLC Supplier No. 250MGHY141371
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromo-PEG2-CH2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.

  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective protein degradation
  • Suitable for laboratory chemical applications
  • Purity of 97.0% (NMR)
  • Appearance: Colorless to light yellow liquid
  • Molecular weight: 283.16
  • Chemical formula: C10H19BrO4
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.