Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bromo-PEG2-CH2-Boc | 1807518-63-3 | 97.0% | C10H19BrO4 | 500 MG
SDP

Catalog No. 5000353107
Encompass_Preferred

Bromo-PEG2-CH2-Boc is a chemical compound frequently utilized as a PEG-based PROTAC linker building block in various research applications. This compound is stable when stored under recommended conditions, and it is strictly intended for research purposes only.

  • CAS number: 1807518-63-3
  • Purity: 97.0% (NMR)
  • Molecular formula: C10H19BrO4
  • Molecular weight: 283.16
  • Appearance: Colorless to light yellow liquid
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Safety: Harmful if swallowed, causes skin and serious eye irritation, may cause respiratory irritation

Catalog No. 50-003-53107 Supplier Medchemexpress LLC Supplier No. HY141371500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromo-PEG2-CH2-Boc is a chemical compound frequently utilized as a PEG-based PROTAC linker building block in various research applications. This compound is stable when stored under recommended conditions, and it is strictly intended for research purposes only.

  • CAS number: 1807518-63-3
  • Purity: 97.0% (NMR)
  • Molecular formula: C10H19BrO4
  • Molecular weight: 283.16
  • Appearance: Colorless to light yellow liquid
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Safety: Harmful if swallowed, causes skin and serious eye irritation, may cause respiratory irritation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.