Learn More
Medchemexpress LLC Bromo-PEG2-phosphonic acid diethyl ester | 1226767-94-7 | C10H22BrO5P | 1 G
SDP
Catalog No. 5000353090
Encompass_Preferred
Bromo-PEG2-phosphonic acid diethyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • A PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-53090 Supplier Medchemexpress LLC Supplier No. HY1412991G
Bromo-PEG2-phosphonic acid diethyl ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • A PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • For research use only