Learn More
Medchemexpress LLC Bromo-PEG2-phosphonic acid diethyl ester | 1226767-94-7 | C10H22BrO5P | 100 MG
SDP
Catalog No. 5000353830
Encompass_Preferred
Bromo-PEG2-phosphonic acid diethyl ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This compound facilitates the creation of bifunctional molecules that leverage the ubiquitin-proteasome system to degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53830 Supplier Medchemexpress LLC Supplier No. HY141299100MG
Bromo-PEG2-phosphonic acid diethyl ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This compound facilitates the creation of bifunctional molecules that leverage the ubiquitin-proteasome system to degrade target proteins.
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins